Merck
[Endoscopic sinus surgery for a paranasal sinuses mucocele with light guide and dacryoendoscopy].
Nihon Jibiinkoka Gakkai kaiho (2014-01-09)
Toshihiko Iwahashi, Takashi Shikina, Masahiro Kawamoto, Ryuichi Mochizuki, Maki Yamashita, Hidenori Inohara
RESUMO
It is hard to cure dacryocystitis caused by a paranasal sinus mucocele with treatment which only targets the mucocele. Also, it is difficult to identify the lacrimal sac and the nasolacrimal duct preoperatively and intraoperatively when the lacrimal passage is markedly changed by the mucocele or previous surgery. We experienced four cases of mucocele complicated by lacrimal stenosis or obstruction. We performed marsupialization of the mucocele and direct silicon intubation or endoscopic dacryocystorhinostomy simultaneously with the use of a fiberoptic illuminator or dacryoendoscopy. Assisted by those devices, lacrimal procedures can now be done quickly and safely regardless of the surgeon's experience. In addition, performing surgeries both for the lacrimal passage and for the mucocele at the same time can minimize the burden on patients.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Silicon, powder, −60 mesh, 99.998% trace metals basis
Sigma-Aldrich
Silicon, pieces, 99.95% trace metals basis
Sigma-Aldrich
Silicon, powder, −325 mesh, 99% trace metals basis
Sigma-Aldrich
Silicon, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.999% trace metals basis
Sigma-Aldrich
Silicon, nanopowder, <100 nm particle size (TEM), ≥98% trace metals basis
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer, <111>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.3 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), contains phosphorus as dopant, <111>, N-type, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.5 mm