  • Search Results
  • Double minutes and MYC amplification: a combined May-Grunwald Giemsa and fluorescence in situ hybridization study.
Double minutes and MYC amplification: a combined May-Grunwald Giemsa and fluorescence in situ hybridization study.
American journal of clinical pathology (2014-01-18)
K F Wong, Lisa L P Siu, W S Wong
ABSTRACT
To report the demonstration of double minutes with MYC amplification in a case of myeloproliferative neoplasm with monocytosis in transformation by a combination of standard karyotyping and interphase and metaphase fluorescence in situ hybridization (FISH). To determine the lineage involvement, we applied combined morphology and an interphase FISH study using dual-color break-apart probes for MYC on peripheral blood film. MYC amplification was demonstrated in both myeloid and monocytic cells but not lymphocytes. The MYC amplification was not associated with loss of MYC signals at the homologous 8q24 regions where the genes were located. Furthermore, the extent of MYC amplification has been shown to diminish as the granulocytes mature. Combined morphology and FISH study has shown a pluripotent myeloid disorder and also an inverse relationship between cell maturity and MYC amplification.
Sigma-Aldrich
Eosin Y disodium salt, certified by the Biological Stain Commission
Sigma-Aldrich
Eosin Y disodium salt, Dye content ≥85 %
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
Eosin Y
Sigma-Aldrich
Eosin Y solution, 5 wt. % in H2O
Sigma-Aldrich
Methylene Blue solution, for microscopy, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology
Sigma-Aldrich
Methylene Blue solution, for microscopy