  • Search Results
  • [Growth characteristics of Fusarium poae (Peck) Wollenw. and Penicillium funiculosum Thom strains].
[Growth characteristics of Fusarium poae (Peck) Wollenw. and Penicillium funiculosum Thom strains].
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993) (2014-02-01)
I N Kurchenko, A K Pavlichenko, E M Iur'eva
ABSTRACT
A comparative study of growth characteristics and peculiarities of glucose utilization by Fusarium poae and Penicillium funiculosum strains isolated from different habitats was conducted. It was found that F. poae and P. funiculosum strains differ as to their growth characteristics. Specific growth rate of endophytic strain F. poae was maximal (0.38 h(-1)), lower for plant pathogenic one (0.30 h(-1)) and the lowest for soil strain (0.18 hr(-1)). In contrast, the level of biomass accumulation was the highest for soil strain F. poae and minimal for plant pathogenic strain, endophytic strain took up an intermediate position. High economic coefficient was characteristic of soil strain and minimal of endophyte at medium biomass level and specific growth rate. In contrast to the strains of F. poae, the specific growth rate of soil strain of P. funiculosum was higher than that of endophyte one (0.35 and 0.24 h(-1), respectively). Biomass level of endophytic strain was significantly higher than that of the soil strain, and economic coefficient was 2.3 times higher for endophyte in comparison with soil strain.
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Supelco
D-(+)-Glucose solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C