• Início
  • Resultados da busca
  • Cr(VI) removal from aqueous solution with bamboo charcoal chemically modified by iron and cobalt with the assistance of microwave.
Cr(VI) removal from aqueous solution with bamboo charcoal chemically modified by iron and cobalt with the assistance of microwave.
Journal of environmental sciences (China) (2014-02-14)
Wei Wang, Xuejiang Wang, Xin Wang, Lianzhen Yang, Zhen Wu, Siqing Xia, Jianfu Zhao
RESUMO
Bamboo charcoal (BC) was used as starting material to prepare Co-Fe binary oxideloaded adsorbent (Co-Fe-MBC) through its impregnation in Co(NO3)2, FeCl3 and HNO3 solutions simultaneously, followed by microwave heating. The low-cost composite was characterized and used as an adsorbent for Cr(VI) removal from water. The results showed that a cobalt and iron binary oxide (CoFe2O4) was uniformly formed on the BC through redox reactions. The composite exhibited higher surface area (331 m2/g) than that of BC or BC loaded with Fe alone (Fe-MBC). The adsorption of Cr(VI) strongly depended on solution pH, temperature and ionic strength. The adsorption isotherms followed the Langmuir isotherm model well, and the maximum adsorption capacities for Cr(VI) at 288 K and pH 5.0 were 35.7 and 51.7 mg/g for Fe-MBC and Co-Fe-MBC, respectively. The adsorption processes were well fitted by the pseudo second-order kinetic model. Thermodynamic parameters showed that the adsorption of Cr(VI) onto both adsorbents was feasible, spontaneous, and exothermic under the studied conditions. The spent Co-Fe-MBC could be readily regenerated for reuse.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, deionized
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Iron, ≥99%, reduced, powder (fine)
Supelco
Water, for TOC analysis
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
Carbonyl iron, ≥97% Fe basis
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Iron, powder, −325 mesh, 97%
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Iron, granular, 10-40 mesh, >99.99% trace metals basis
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis