Merck
Ecotoxicity of pristine graphene to marine organisms.
Ecotoxicology and environmental safety (2014-02-11)
Carlo Pretti, Matteo Oliva, Roberta Di Pietro, Gianfranca Monni, Giorgio Cevasco, Federica Chiellini, Christian Pomelli, Cinzia Chiappe
ABSTRACT
The ecotoxicity of pristine graphene nanoparticles (GNC1, PGMF) in model marine organisms was investigated. PGMF resulted more toxic than GNC1 to the bioluminescent bacterium Vibrio fischeri and the unicellular alga Dunaliella tertiolecta on the basis of EC50 values (end-points: inhibition of bioluminescence and growth, respectively). No acute toxicity was demonstrated with respect to the crustacean Artemia salina although light microscope images showed the presence of PGMF and GNC1 aggregates into the gut; a 48-h exposure experiment revealed an altered pattern of oxidative stress biomarkers, resulting in a significant increase of catalase activities in both PGMF and GNC1 1mg/L treated A. salina and a significant increase of glutathione peroxidase activities in PGMF (0.1 and 1mg/L) treated A. salina. Increased levels of lipid peroxidation of membranes was also observed in PGMF 1mg/L exposed A. salina.
Sigma-Aldrich
Catalase from bovine liver, ≥20000 units/mg protein, lyophilized powder
Sigma-Aldrich
Catalase from Aspergillus niger, ammonium sulfate suspension, ≥4,000 units/mg protein
Sigma-Aldrich
Glutathione Peroxidase from bovine erythrocytes, lyophilized powder, ≥300 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, ≥10,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 10,000-40,000 units/mg protein
Sigma-Aldrich
Catalase from human erythrocytes, ≥90% (SDS-PAGE), buffered aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous solution, ≥30,000 units/mg protein