Merck
Accumulation and toxicological response of atrazine in rice crops.
Ecotoxicology and environmental safety (2014-02-18)
Jia Jun Zhang, Yi Chen Lu, Jin Jin Zhang, Li Rong Tan, Hong Yang
ABSTRACT
Atrazine is one of the most widely used herbicides for controlling weeds and grasses. Due to its intensive use, it has become a serious contaminant in soil and water. To evaluate impact of atrazine on graminaceous crops, experiments focusing on atrazine accumulation and toxic response in rice (Oryza sativa) were carried out. Treatment with atrazine at 0.05-0.8 mg L(-1) for 6 d reduced elongation of shoot and root. Compared with a mock treatment, the elongation of shoot with atrazine was 67.1 percent of the control, whereas that of root was 79.5 percent, indicating that the shoot was more affected than the root. Atrazine was readily absorbed by rice from media. Although the quantitative absorption of atrazine was positively correlated with the external supply of the herbicide, translocation of atrazine from roots to the above-ground was reduced from 39.88±6.26 (at 0.05 mg L(-1)) to 9.25±0.27 (0.8 mg L(-1)). While accumulation of atrazine in rice plants led to toxic responses such as over-generation of hydrogen peroxide and superoxide anions, it triggered the plant defense system against the herbicide-induced oxidative stress. This was best presented by the enhanced activities of several antioxidant enzymes (e.g. superoxide dismutase, catalase and peroxidase) and expression of genes responsible for the tolerance to atrazine toxicity.
Sigma-Aldrich
Catalase from Aspergillus niger, ammonium sulfate suspension, ≥4,000 units/mg protein
Sigma-Aldrich
Catalase from human erythrocytes, ≥90% (SDS-PAGE), buffered aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 10,000-40,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein
Supelco
Atrazine, PESTANAL®, analytical standard
Sigma-Aldrich
Catalase from bovine liver, ≥20000 units/mg protein, lyophilized powder
Supelco
Atrazine, certified reference material, TraceCERT®
Supelco
Atrazine, analytical standard
Supelco
Atrazine solution, 1000 μg/mL in methyl tert-butyl ether, certified reference material
Sigma-Aldrich
Catalase from bovine liver, aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, ≥10,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)