  • Search Results
  • Protective effect of secondary plant metabolites from Ipomoea aquatica Forsk. against carbofuran induced damages.
Protective effect of secondary plant metabolites from Ipomoea aquatica Forsk. against carbofuran induced damages.
Indian journal of experimental biology (2014-03-04)
Sanjukta Datta, Mahuya Sinha, Dipesh Das, Santinath Ghosh, Pubali Dhar
ABSTRACT
Plausible interactions between food contaminants and natural constituents in vivo and protective effect of polyphenols present in I. aquatica against carbofuran toxicity in Charles Foster rats were evaluated. Determinations based on antioxidant enzyme activities showed significant alterations in glutathione, glutathione peroxidase, superoxide dismutase and catalase in tissues (liver and brain) and plasma of pesticide treated group while polyphenolic extracts from I. aquatica (IAE) attenuated their activities when given alongwith carbofuran. IAE decreased enhanced lipid peroxidation levels in plasma and erythrocyte membrane and cholesterol levels in brain and plasma. IAE also minimized histopathological degenerative changes produced by carbofuran. While single cell gel electrophoresis showed that secondary metabolites in leafy vegetables produced a combinatorial effect with pesticide at cellular level, DNA fragmentation level in bone marrow cells showed a decline in the IAE treated rats. Food safety adversely affected by various chemical contaminants can be retained by plant polyphenols and secondary plant constituents that can be found together in bolus. Therefore, the present study gives an insight into the protective role of naturally found polyphenols against pesticide toxicity.
Sigma-Aldrich
Superoxide Dismutase from bovine erythrocytes, lyophilized powder, ≥3,000 units/mg protein
Sigma-Aldrich
Superoxide Dismutase from bovine liver, lyophilized powder, ≥1500 units/mg protein
Sigma-Aldrich
Superoxide Dismutase from bovine erythrocytes, BioUltra, lyophilized powder, ≥4,500 units/mg protein, ≥97% (SDS-PAGE)
Sigma-Aldrich
Superoxide Dismutase from bovine liver, ammonium sulfate suspension, 2,000-6,000 units/mg protein (biuret)
Sigma-Aldrich
Superoxide Dismutase from bovine erythrocytes, ≥3,000 units/mg protein, BioReagent, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Superoxide Dismutase bovine, recombinant, expressed in E. coli, lyophilized powder, ≥2500 units/mg protein, ≥90% (SDS-PAGE)
Sigma-Aldrich
Superoxide Dismutase I human, recombinant, expressed in E. coli
Sigma-Aldrich
Catalase from Aspergillus niger, ammonium sulfate suspension, ≥4,000 units/mg protein
Sigma-Aldrich
Glutathione Peroxidase from bovine erythrocytes, lyophilized powder, ≥300 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, ≥10,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 10,000-40,000 units/mg protein
Sigma-Aldrich
Catalase from human erythrocytes, ≥90% (SDS-PAGE), buffered aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Superoxide Dismutase from human erythrocytes, essentially salt-free, lyophilized powder, ≥2,500 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, ≥20000 units/mg protein, lyophilized powder