  • Search Results
  • [Cytoplasmic superoxide dismutase activity is a sensitive indicator of the antioxidant status of the rat liver and brain].
[Cytoplasmic superoxide dismutase activity is a sensitive indicator of the antioxidant status of the rat liver and brain].
Biomeditsinskaia khimiia (2014-04-23)
T V Sirota, M V Zakharchenko, M N Kondrashova
ABSTRACT
Several parameters of the cytoplasmic enzymatic antioxidant system of the liver and brain of the rat have been investigated under conditions of immobilization stress and of an antioxidant preparation in the diet of animals. These included superoxide dismutase (SOD) and glutathione reductase (GR) activities and nonspecific NADPH oxidation. Only changes in the activity of SOD both in the liver and brain were revealed. In the liver of animals that receive no preparation, a decrease in the activity of SOD after 30-min immobilization and its restoration after a 360-min immobilization were observed. In the brain, the activity of SOD decreased only in preconditioned animals after 30 and 360 min of exposure to stress. In addition, the activity of SOD in the brain of preconditioned animals, both stressed and unstressed, was lower than in the corresponding groups of control animals. It is probable that, under the conditions of immobilization stress, the level of reactive oxygen species (ROS) and as a consequence the activity of SOD decrease. The intake of an antioxidant preparation under these conditions seems to be not correct.
Sigma-Aldrich
Catalase from Aspergillus niger, ammonium sulfate suspension, ≥4,000 units/mg protein
Sigma-Aldrich
Catalase from human erythrocytes, ≥90% (SDS-PAGE), buffered aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 10,000-40,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein
Sigma-Aldrich
Glutathione Peroxidase from bovine erythrocytes, lyophilized powder, ≥300 units/mg protein
Sigma-Aldrich
Superoxide Dismutase from human erythrocytes, essentially salt-free, lyophilized powder, ≥2,500 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, ≥20000 units/mg protein, lyophilized powder
Sigma-Aldrich
Catalase from bovine liver, aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, ≥10,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)
Sigma-Aldrich
Superoxide Dismutase from bovine liver, lyophilized powder, ≥1500 units/mg protein
Sigma-Aldrich
Superoxide Dismutase from bovine erythrocytes, ≥3,000 units/mg protein, BioReagent, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Superoxide Dismutase from bovine erythrocytes, BioUltra, lyophilized powder, ≥4,500 units/mg protein, ≥97% (SDS-PAGE)
Sigma-Aldrich
Superoxide Dismutase from bovine liver, ammonium sulfate suspension, 2,000-6,000 units/mg protein (biuret)
Sigma-Aldrich
Superoxide Dismutase from bovine erythrocytes, lyophilized powder, ≥3,000 units/mg protein
Sigma-Aldrich
Superoxide Dismutase bovine, recombinant, expressed in E. coli, lyophilized powder, ≥2500 units/mg protein, ≥90% (SDS-PAGE)
Sigma-Aldrich
Superoxide Dismutase I human, recombinant, expressed in E. coli