  • Search Results
  • [Blood oxygen transport, prooxidant -- antioxidant status, and vasoactive characteristics of vascular endothelium in rats treated with endotoxin and taurine].
[Blood oxygen transport, prooxidant -- antioxidant status, and vasoactive characteristics of vascular endothelium in rats treated with endotoxin and taurine].
Eksperimental'naia i klinicheskaia farmakologiia (2014-05-08)
T S Milosh, N E Maksimovich
ABSTRACT
Experiments on a group of 74 pregnant rats upon intramuscular introduction of E. coli lipopolysaccharides during pregnancy revealed the correction effect of taurine on the blood oxygen transport function, prooxidant - antioxidant status, and vasoactive characteristics of vascular endothelium.
Sigma-Aldrich
Catalase from Aspergillus niger, ammonium sulfate suspension, ≥4,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 10,000-40,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, ≥10,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from human erythrocytes, ≥90% (SDS-PAGE), buffered aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, ≥20000 units/mg protein, lyophilized powder
Sigma-Aldrich
Catalase−Agarose, pH 5.4(isoelectric point), ammonium sulfate suspension, enzyme from bovine liver