  • Search Results
  • Non-topography-guided photorefractive keratectomy for the correction of residual mild refractive errors after ICRS implantation and CXL in keratoconus.
Non-topography-guided photorefractive keratectomy for the correction of residual mild refractive errors after ICRS implantation and CXL in keratoconus.
Journal of refractive surgery (Thorofare, N.J. : 1995) (2014-04-08)
Ali Dirani, Ali Fadlallah, Zeba A Syed, Elias Chelala, Ziad Khoueir, George Cherfan, Elias Jarade
ABSTRACT
To evaluate the safety and clinical outcomes of non-topography-guided photorefractive keratectomy (PRK) for the treatment of residual mild refractive errors 6 months after sequential intracorneal ring segment (ICRS) implantation and corneal collagen cross-linking (CXL) in stable keratoconus. This retrospective study included 17 eyes of 14 patients with mild to moderate keratoconus. The ICRS implantation and CXL were performed sequentially with a 4-week interval and non-topography-guided PRK was performed at least 6 months after CXL. Data were collected preoperatively and at the 6-month follow-up visits. ICRS implantation and CXL induced a significant decrease in keratometry and refraction and an increase in visual acuity. At the 6-month follow-up after ICRS implantation and CXL, uncorrected and corrected distance visual acuity (UDVA and CDVA) significantly improved from 1.17 ± 0.38 and 0.44 ± 0.09 logMAR preoperatively to 0.45 ± 0.11 and 0.17 ± 0.08 logMAR (P = .001) postoperatively, respectively. The mean spherical error decreased from -5.45 ± 1.64 to -2.57 ± 1.15 D (P = .01) and the mean cylinder from 3.86 ± 1.15 to 2.13 ± 1.11 D (P = .01). At the 6-month follow-up after PRK, UDVA significantly improved to 0.18 ± 0.06 logMAR and CDVA was 0.15 ± 0.05 logMAR. The mean spherical error and mean cylinder significantly decreased to -1.10 ± 0.41 D (P = .02) and 0.98 ± 0.37 D (P = .046), respectively. No intraoperative or postoperative complications occurred. At the 6-month follow-up, non-topography-guided PRK after ICRS implantation and CXL was found to be an effective and safe option for correcting residual refractive error and improving visual acuity in patients with moderate keratoconus.
Product Description
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, average Mw 97,000 (Typical), average Mn 46,000 (Typical)
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 50,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 10,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 100,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 2,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 20,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 8,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 2,480,000
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
Poly(methyl methacrylate)
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 4,000
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~350,000 by GPC
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~15,000 by GPC, powder
Riboflavin for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Poly(methyl methacrylate), isotactic, >80% isotactic
Supelco
Riboflavin (B2), analytical standard