  • Search Results
  • Cationic Poly-L-Lysine-Fe2O3/SiO2 nanoparticles loaded with small interference RNA: Application to silencing gene expression in primary rat neurons.
Cationic Poly-L-Lysine-Fe2O3/SiO2 nanoparticles loaded with small interference RNA: Application to silencing gene expression in primary rat neurons.
Journal of nanoscience and nanotechnology (2014-04-17)
Xuewen Cui, Ruijiang Liu, Ziwen Liu, Xiangqian Shen, Qiuju Wang, Xiaoli Tan
ABSTRACT
The cationic PLL-Fe2o3/SiO2-siRNA nanoparticles with an average crystallite size of about 20 nm were prepared and delivered into primary rat neurons for knockdown of gene expression. Primary rat fetal neurons were scratched to simulate the injury of central nervous system and then were transfected with PLL-Fe2O3/SiO2-siRNA nanoparticles. Optical microscopy, fluorescence microscopy, Western blotting, immunofluorescence, and MTT assays were employed to test the cytotoxicity, the efficiency of encapsulation and targeted gene silencing. The results indicated that the PLL-Fe2O3/SiO2-siRNA nanoparticles have a remarkable efficiency of encapsulation and targeted gene silencing with negligible cytotoxicity. It could be concluded that the PLL-Fe2O3/SiO2 nanoparticles are a promising delivery carrier of siRNA.
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Iron(III) oxide, dispersion, nanoparticles, ≤110 nm particle size, 15 wt. % in ethanol
Sigma-Aldrich
Silica, nanopowder, spec. surface area 175-225 m2/g (BET), 99.8% trace metals basis
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
Silica
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
Sigma-Aldrich
Iron(III) oxide, nanopowder, <50 nm particle size (BET)
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Iron(III) oxide, powder, <5 μm, ≥96%