  • Search Results
  • Bioavailability and biological activity of L-ascorbic acid 2-O-alpha-glucoside.
Bioavailability and biological activity of L-ascorbic acid 2-O-alpha-glucoside.
Journal of nutritional science and vitaminology (1992-01-01)
I Yamamoto, N Muto
ABSTRACT
AA-2G is a new stable derivative of AsA which is efficiently synthesized by regioselective transglucosylation with alpha-glucosidase and CGTase. AA-2G serves as a vitamin C supplement in experimental animals. AA-2G is easily hydrolyzed in vivo by alpha-glucosidase and also synthesized as a metabolite under some specified conditions. AA-2G stimulates collagen synthesis in cultured fibroblasts and enhances antibody production in cultured splenocytes. AA-2G which has no cytotoxicity is a promising AsA derivative for medical and nutritional uses.
Product Description
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®