Possible role for dietary lutein and zeaxanthin in visual development.
Nutrition reviews (2008-11-21)
Billy R Hammond
RESUMO
The possibility that the macular carotenoids, lutein (L), and zeaxanthin (Z), could retard age-related changes in the eye and prevent the eye diseases that result from such changes (namely, cataract and macular degeneration) has been carefully studied. A role for the carotenoids very early in life, however, has received far less attention. Nevertheless, an influence on visual development is likely. Retinal L and Z, for instance, would influence the development of the visual system if they 1) altered input during a critical/sensitive period of visual development and/or 2) influenced maturation and/or 3) protected the retina during a period when it was particularly vulnerable. The available evidence indicates that the pigments may play a role in all three of these areas.
MATERIAIS
Número do produto
Marca
Descrição do produto
Supelco
Lutein, analytical standard
Supelco
Zeaxanthin, analytical standard
Sigma-Aldrich
Xanthophyll, from marigold