  • Search Results
  • Cerebral glucose metabolism in preclinical and prodromal Alzheimer's disease.
Cerebral glucose metabolism in preclinical and prodromal Alzheimer's disease.
Expert review of neurotherapeutics (2010-10-28)
Karl Herholz
ABSTRACT
Assessment of regional cerebral glucose metabolism by (18)F-2-fluoro-2-deoxy-D-glucose (FDG)-PET at resting state is a standard functional technique to assess cerebral function. Group studies identified significant regional metabolic impairment in asymptomatic individuals at increased risk for dementia. Substantial impairment of FDG uptake in temporoparietal association cortices emerges as a reliable predictor of rapid progression to dementia in mild cognitive impairment patients and could, therefore, serve as a biomarker for the diagnosis of prodromal Alzheimer's disease. Frontal and temporoparietal metabolic impairment is closely related to progression of disease in longitudinal studies, and multicenter studies suggest its utility as an outcome parameter to increase the efficiency of therapeutic trials.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Supelco
D-(+)-Glucose solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous