Formation of ring-opened and rearranged products of guanine: mechanisms and biological significance.
Free radical biology & medicine (2012-05-16)
N R Jena, P C Mishra
RESUMO
DNA damage by endogenous and exogenous agents is a serious concern, as the damaged products can affect genome integrity severely. Damage to DNA may arise from various factors such as DNA base modifications, strand break, inter- and intrastrand crosslinks, and DNA-protein crosslinks. Among these factors, DNA base modification is a common and important form of DNA damage that has been implicated in mutagenesis, carcinogenesis, and many other pathological conditions. Among the four DNA bases, guanine (G) has the smallest oxidation potential, because of which it is frequently modified by reactive species, giving rise to a plethora of lethal lesions. Similarly, 8-oxo-7,8-dihydroguanine (8-oxoG), an oxidatively damaged guanine lesion, also undergoes various degradation reactions giving rise to several mutagenic species. The various products formed from reactions of G or 8-oxoG with different reactive species are mainly 2,6-diamino-4-oxo-5-formamidopyrimidine, 2,5-diamino-4H-imidazolone, 2,2,4-triamino-5-(2H)-oxazolone, 5-guanidino-4-nitroimidazole, guanidinohydantoin, spiroiminodihydantoin, cyanuric acid, parabanic acid, oxaluric acid, and urea, among others. These products are formed from either ring opening or ring opening and subsequent rearrangement. The main aim of this review is to provide a comprehensive overview of various possible reactions and the mechanisms involved, after which these ring-opened and rearranged products of guanine would be formed in DNA. The biological significance of oxidatively damaged products of G is also discussed.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Guanine, 98%
Supelco
Guanine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Guanine, BioUltra