Número do produto
Marca
Descrição do produto
Silicon, rod, 50mm, diameter 6mm, single crystal, -111, 99.999%
Silicon, rod, 50mm, diameter 3.15mm, single crystal - random orientation, 100%
Silicon, rod, 10mm, diameter 2.0mm, crystalline, 100%
Silicon, rod, 50mm, diameter 5.0mm, crystalline, 100%
Silicon, rod, 50mm, diameter 5mm, single crystal, -111, 99.999%
Silicon, rod, 80mm, diameter 20mm, single crystal, -100, 99.999%
Silicon, rod, 25mm, diameter 3.15mm, single crystal - random orientation, 100%
Silicon, disks, 15.9mm, thickness 0.38mm, single crystal, n-type, 100%
Silicon, single crystal, -111, n-type, phosphorus doped, diameter 76.2mm, length 0.75mm, 100%
Silicon, sphere, 50pcs, diameter 2.0mm, precision sphere grade 25
Silicon, sheet, 10x10mm, thickness 0.5mm, single crystal, -100, 100%
Silicon, rod, 40mm, diameter 20mm, single crystal, -100, 99.999%
Silicon, rod, 100mm, diameter 5.0mm, crystalline, 100%
Silicon, rod, 100mm, diameter 2.0mm, crystalline, 100%
Sigma-Aldrich
Silicon, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.999% trace metals basis
Silicon, rod, 50mm, diameter 2.0mm, crystalline, 100%
Silicon, sheet, 25x25mm, thickness 1.0mm, single crystal, -100, 100%
Silicon, sheet, 14x14mm, thickness 1.0mm, single crystal, -111, 100%
Silicon, rod, 50mm, diameter 5mm, single crystal, -100, 99.999%
Silicon, sheet, 25x25mm, thickness 1.0mm, polycrystalline, 99.999%
Silicon, sheet, 10x10mm, thickness 0.6mm, single crystal, -100, 100%
Silicon, sheet, 14x14mm, thickness 1.0mm, single crystal, -100, 100%
Silicon, sheet, 40x40mm, thickness 3.0mm, single crystal, p-type, 100%
Silicon, rod, 100mm, diameter 25mm, crystalline, 100%
Silicon, disks, 13mm, thickness 0.38mm, single crystal, 100%
Silicon, disks, 15.9mm, thickness 0.38mm, single crystal, n-type, 100%
Silicon, rod, 100mm, diameter 10.0mm, single crystal - random orientation, 100%
Silicon, sheet, 52x52mm, thickness 1.0mm, polycrystalline, 99.999%
Silicon, sphere, 10pcs, diameter 2.0mm, precision sphere grade 25
Silicon, rod, 100mm, diameter 12.7mm, single crystal - random orientation, 100%