  • Search Results
  • Child neurology: differential diagnosis of a low CSF glucose in children and young adults.
Child neurology: differential diagnosis of a low CSF glucose in children and young adults.
Neurology (2013-12-11)
Wilhelmina G Leen, Cornelis J de Wit, Ron A Wevers, Baziel G van Engelen, Erik-Jan Kamsteeg, Joerg Klepper, Marcel M Verbeek, Michèl A Willemsen
ABSTRACT
Analysis of CSF is daily routine in patients with acute neurologic disorders like CNS infections. In those patients, the finding of a low CSF glucose may influence further diagnostic workup and therapeutic choices. The interpretation of a low CSF glucose in patients with a chronic neurologic disorder, however, is a less common practice. We present a practical overview on the differential diagnosis of a low CSF glucose and stress the importance of recognizing a low CSF glucose as the diagnostic marker for GLUT1 deficiency syndrome, a treatable neurometabolic disorder.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Supelco
D-(+)-Glucose solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material