Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Chromium, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 99.99+%
Chromium, foil, light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 99.99+%
Chromium, microfoil, disks, 10mm, thinness 1.0μm, specific density 719μg/cm2, permanent mylar 3.5μm support, 99.99+%
Chromium, foil, not light tested, 50x50mm, thickness 0.004mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 99.99+%
Chromium, microfoil, disks, 25mm, thinness 0.5μm, specific density 355μg/cm2, permanent mylar 3.5μm support, 99.99+%
Chromium, foil, not light tested, 50x50mm, thickness 0.02mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 99.99+%
Chromium, microfoil, disks, 25mm, thinness 0.05μm, specific density 37.27μg/cm2, permanent mylar 3.5μm support, 99.99+%
Chromium, rod, 50mm, diameter 5.0mm, 99.7+%
Chromium, foil, not light tested, 50x50mm, thickness 0.005mm, permanent polyester support, 99.99+%
Chromium, foil, light tested, 25x25mm, thickness 0.075mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 50x50mm, thickness 0.03mm, permanent polyester support, 99.99+%
Chromium, microfoil, disks, 10mm, thinness 0.05μm, specific density 37.27μg/cm2, permanent mylar 3.5μm support, 99.99+%
Chromium, foil, not light tested, 50x50mm, thickness 0.0025mm, permanent polyester support, 99.99+%
Chromium, foil, 100x100mm, thickness 3.0mm, hard, 99.95%
Chromium, foil, light tested, 25x25mm, thickness 0.0125mm, permanent polyester support, 99.99+%
Chromium, foil, light tested, 25x25mm, thickness 0.008mm, permanent polyester support, 99.99+%
Chromium, foil, light tested, 25x25mm, thickness 0.03mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 50x50mm, thickness 0.006mm, permanent polyester support, 99.99+%
Chromium, foil, light tested, 25x25mm, thickness 0.04mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 99.99+%
Chromium, foil, 50x50mm, thickness 3.0mm, hard, 99.95%