• Início
  • Resultados da busca
  • A facile approach for the polymer grafting of silica based on tandem reversible addition fragmentation chain transfer/click chemistry and its application in high performance liquid chromatography.
A facile approach for the polymer grafting of silica based on tandem reversible addition fragmentation chain transfer/click chemistry and its application in high performance liquid chromatography.
Journal of chromatography. A (2014-06-10)
Zheng Zhang, Ming-Luan Chen, Xiao-Dong Cheng, Zhi-Guo Shi, Bi-Feng Yuan, Yu-Qi Feng
RESUMO
In this work, a tandem reversible addition fragmentation chain transfer (RAFT)/click chemistry method was developed to prepare amide-polystyrene-silica (NHCO-Psty-silica) stationary phase. Styrene was immobilized on amino-silica surface via an azide functionalized RAFT agent in a one-pot procedure. The resultant NHCO-Psty-silica column demonstrates better performance for shielding of residue silanols than traditional ODS column, which was ascertained by Engelhardt test (E test), Tanaka test (T test), Galushko test (G test), and Walters test (W test). Our results showed lower values of silanol activity calculated according to the formula in these standard tests for NHCO-Psty-silica column compared to the ODS column we tested. As a result, the NHCO-Psty-silica is suitable for the separation of basic compounds. The hydrophobic, anion exchanging and π-π interaction of the column toward analytes were also evaluated. Moreover, the NHCO-Psty-silica column also showed excellent stability with pure water as mobile phase.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica, nanopowder, spec. surface area 175-225 m2/g (BET), 99.8% trace metals basis
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Sigma-Aldrich
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O