  • Search Results
  • Effects of dietary glucose and fructose upon cannabinoid CB1 receptor functionality in the rat brain: a pilot study.
Effects of dietary glucose and fructose upon cannabinoid CB1 receptor functionality in the rat brain: a pilot study.
Life sciences (2014-06-15)
Maria Luisa Rojo, Mariateresa Cipriano, Ingegerd Söderström, Kotryna Simonytė, Tommy Olsson, Christopher J Fowler
ABSTRACT
A high consumption of fructose leads not only to peripheral changes in insulin sensitivity and vascular function, but also to central changes in several brain regions. Given the role of the endogenous cannabinoid system in the control of energy intake, we undertook a pilot study to determine whether a high fructose diet produced changes in brain CB1 receptor functionality. Male rats given access ad libitum to normal chow were given either water, glucose or fructose solutions to drink. CB1 receptor functionality was measured autoradiographically as the increase in [(35)S]GTPγS binding produced by the agonist CP55,940. Seven regions were investigated: the prefrontal cortex, caudate-putamen, hippocampal CA1-CA3, dentate gyrus, amygdala, and dorsomedial and ventromedial hypothalami. Two-way robust Wilcoxon analyses for each brain region indicated that the dietary treatment did not produce significant main effects upon agonist-stimulated [(35)S]GTPγS binding in any of the regions, in contrast to a significant main effect upon both leptin and adiponectin levels in the blood. However, a MANCOVA of the data controlling for leptin and adiponectin as co-variables identified a significant effect of glucose and fructose treatment for five weeks upon the [(35)S]GTPγS response in the ventromedial hypothalamus, a region of importance for regulation of appetite. It is concluded from this pilot study that palatable solutions do not produce overt changes in brain CB1 receptor functionality, although subtle changes in discrete brain regions may occur.
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Supelco
D-(+)-Glucose solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material