Merck
Synthetic biology. Programmable on-chip DNA compartments as artificial cells.
Science (New York, N.Y.) (2014-08-16)
Eyal Karzbrun, Alexandra M Tayar, Vincent Noireaux, Roy H Bar-Ziv
RESUMO
The assembly of artificial cells capable of executing synthetic DNA programs has been an important goal for basic research and biotechnology. We assembled two-dimensional DNA compartments fabricated in silicon as artificial cells capable of metabolism, programmable protein synthesis, and communication. Metabolism is maintained by continuous diffusion of nutrients and products through a thin capillary, connecting protein synthesis in the DNA compartment with the environment. We programmed protein expression cycles, autoregulated protein levels, and a signaling expression gradient, equivalent to a morphogen, in an array of interconnected compartments at the scale of an embryo. Gene expression in the DNA compartment reveals a rich, dynamic system that is controlled by geometry, offering a means for studying biological networks outside a living cell.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Silicon, powder, −60 mesh, 99.998% trace metals basis
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Sigma-Aldrich
Silicon, powder, −325 mesh, 99% trace metals basis
Sigma-Aldrich
Silicon, pieces, 99.95% trace metals basis
Sigma-Aldrich
Silicon, wafer, <111>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.3 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Silicon, sphere, 20pcs, diameter 2.0mm, precision sphere grade 25
Silicon, rod, 50mm, diameter 3.15mm, single crystal - random orientation, 100%
Silicon, rod, 80mm, diameter 20mm, single crystal, -100, 99.999%
Silicon, single crystal, -111, n-type, phosphorus doped, diameter 76.2mm, length 0.75mm, 100%
Silicon, rod, 50mm, diameter 5mm, single crystal, -100, 99.999%
Silicon, sheet, 40x40mm, thickness 3.0mm, single crystal, p-type, 100%
Silicon, sphere, 50pcs, diameter 2.0mm, precision sphere grade 25
Silicon, rod, 100mm, diameter 12.7mm, single crystal - random orientation, 100%
Silicon, rod, 50mm, diameter 5.0mm, crystalline, 100%
Silicon, rod, 50mm, diameter 2.0mm, crystalline, 100%
Silicon, rod, 100mm, diameter 5.0mm, crystalline, 100%
Silicon, rod, 25mm, diameter 3.15mm, single crystal - random orientation, 100%
Silicon, sheet, 10x10mm, thickness 0.5mm, single crystal, -100, 100%
Silicon, sheet, 10x10mm, thickness 0.6mm, single crystal, -100, 100%
Silicon, sphere, 10pcs, diameter 2.0mm, precision sphere grade 25
Silicon, sheet, 25x25mm, thickness 1.0mm, single crystal, -100, 100%
Silicon, rod, 25mm, diameter 2.0mm, crystalline, 100%
Silicon, rod, 50mm, diameter 5mm, single crystal, -111, 99.999%
Silicon, rod, 40mm, diameter 20mm, single crystal, -100, 99.999%
Silicon, rod, 10mm, diameter 2.0mm, crystalline, 100%
Silicon, sheet, 52x52mm, thickness 1.0mm, polycrystalline, 99.999%
Silicon, rod, 100mm, diameter 25mm, crystalline, 100%
Silicon, sheet, 25x25mm, thickness 1.0mm, polycrystalline, 99.999%