Natural attenuation of pharmaceuticals and an illicit drug in a laboratory column experiment.
Chemosphere (2014-03-07)
Andrew M Greenhagen, Melissa E Lenczewski, Monica Carroll
RESUMO
Trace amounts of pharmaceutical compounds have been detected in waters across the United States. Many compounds are released as the result of human ingestion and subsequent excretion of over-the-counter and prescription medications, and illicit drugs. This research utilized columns (30×30cm) of sand and undisturbed fine-grained sediments to simulate injection of wastewater containing pharmaceuticals and an illicit drug, such as would be found in a septic system, leaky sewer, or landfill. The columns were placed in a temperature-controlled laboratory and each was injected with natural groundwater containing known concentrations of caffeine, methamphetamine, and acetaminophen. Natural attenuation of each chemical was observed in all columns. The highest amount removed (approximately 90%) occurred in the undisturbed column injected with methamphetamine, compared with little reduction in the sand column. When the suite of drugs was injected, loss of methamphetamine was less than when methamphetamine was injected alone. The subsurface sediments exhibit the ability to remove a substantial amount of the injected pharmaceuticals and illicit drug; however, complete removal was not achieved. There was little attenuation of injected pharmaceuticals in the sand column which demonstrates a concern for water quality in the environment if pharmaceuticals were to contaminate a sandy aquifer. Understanding the transport of pharmaceuticals in the subsurface environment is an important component of protecting drinking water supplies from contamination.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Supelco
Caffeine, certified reference material, TraceCERT®
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica, nanopowder, spec. surface area 175-225 m2/g (BET), 99.8% trace metals basis
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Supelco
Caffeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Caffeine, BioXtra