• Início
  • Resultados da busca
  • Bone response from a dynamic stimulus on a one-piece and multi-piece implant abutment and crown by finite element analysis.
Bone response from a dynamic stimulus on a one-piece and multi-piece implant abutment and crown by finite element analysis.
The Journal of oral implantology (2013-01-16)
Habib Hajimiragha, Mohammadreza Abolbashari, Saeed Nokar, AmirHossein Abolbashari, Mehrdad Abolbashari
RESUMO
The present study was done to evaluate the effects of different types of abutments on the rate and distribution of stress on the bone surrounding the implant by dynamic finite element analysis method. In this study two ITI abutment models-one-piece and multi-piece-along with fixture, bone, and superstructure have been simulated with the help of company-made models. The maximum Von Mises stress (MVMS) was observed in the distobuccal area of the cortical bone near the crest of implant in two implant models. In the multi-piece abutment, MVMS was higher than the one-piece model (27.9 MPa and 23.3 MPa, respectively). Based on the results of this study, it can be concluded that type of abutment influences the stress distribution in the area surrounding the implant during dynamic loading.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, powder, <45 μm avg. part. size, 99.98% trace metals basis
Sigma-Aldrich
Titanium, powder, −100 mesh, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, sponge, 3-19 mm, 99.5% trace metals basis
Sigma-Aldrich
Titanium, nanoparticles, dispersion, <100 nm particle size, in mineral oil, 98.5% trace metals basis
Sigma-Aldrich
Titanium, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.995% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, crystalline, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 2.0 mm, 99.99% trace metals basis
Titanium, rod, 100mm, diameter 10mm, annealed, 99.6+%
Titanium, tube, 200mm, outside diameter 0.51mm, inside diameter 0.35mm, wall thickness 0.08mm, annealed, 99.6+%
Sigma-Aldrich
Titanium, wire, diam. 0.127 mm, 99.99% trace metals basis
Titanium, rod, 100mm, diameter 2mm, annealed, 99.6+%
Titanium, rod, 500mm, diameter 2mm, as drawn, 99.99+%
Titanium, wire reel, 1m, diameter 0.25mm, as drawn, 99.6+%
Titanium, wire reel, 1m, diameter 0.5mm, as drawn, 99.6+%
Titanium, tube, 500mm, outside diameter 0.51mm, inside diameter 0.35mm, wall thickness 0.08mm, annealed, 99.6+%
Titanium, tube, 500mm, outside diameter 0.81mm, inside diameter 0.61mm, wall thickness 0.1mm, as drawn, 99.6+%
Titanium, tube, 500mm, outside diameter 1.05mm, inside diameter 0.75mm, wall thickness 0.15mm, annealed, 99.6+%
Titanium, tube, 500mm, outside diameter 1.6mm, inside diameter 1.2mm, wall thickness 0.2mm, hard, 99.6+%