• Início
  • Resultados da busca
  • Separation of enantiomers on chiral stationary phase based on chicken α₁-acid glycoprotein: effect of silica particle diameters on column performance.
Separation of enantiomers on chiral stationary phase based on chicken α₁-acid glycoprotein: effect of silica particle diameters on column performance.
Journal of chromatography. A (2014-07-22)
Hisami Matsunaga, Jun Haginaka
RESUMO
The effects of silica particle diameters on performances of chicken α₁-acid glycoprotein (c-AGP)-immobilized silica particle columns were investigated. c-AGP was immobilized onto aminopropyl silica particles, whose nominal particle diameters were 5, 3 and 2.1 μm, activated with N,N'-disuccinimidyl carbonate. The retention factor (k), enantioseparation factor (α), resolution (Rs) and height equivalent to a theoretical plate (H) of solutes on three c-AGP columns were evaluated using a mixture of phosphate buffer and organic modifier as a mobile phase in LC. There were not so much differences in their k and α values among three c-AGP columns, while their Rs values were in the order of 2.1 μm>3 μm>5 μm silica particles and their H values were in the reversed order. Since three c-AGP columns gave almost the same enantioseparation factors for solutes, their highest Rs and lowest H values on a c-AGP-immobilized column prepared with 2.1-μm silica particles came from its highest column efficiency among there c-AGP columns. These results suggest that 2.1-μm silica particles could be useful for the preparation of c-AGP- or protein-based CSPs.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica, nanopowder, spec. surface area 175-225 m2/g (BET), 99.8% trace metals basis
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Sigma-Aldrich
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O