Study of penetration behavior of PCB-DNAPL in a sand layer by a column experiment.
Chemosphere (2014-08-13)
Nobuyasu Okuda, Takaaki Shimizu, Masaru Muratani, Akihiko Terada, Masaaki Hosomi
RESUMO
To better understand the infiltration performances of high concentration PCB oils (KC-300 and KC-1000 polychlorinated biphenyl (PCB) mixtures), representative dense non-aqueous phase liquid (DNAPL), under both saturated and unsaturated conditions, we conducted experiments on a sand column filled with Toyoura Standard Sand. When PCB oil with the volume comparable to the total porosity in the column was supplied, the residual PCB concentrations under PCB-water conditions were 4.9×10(4)mgkg(-1) in KC-300 and 3.9×10(4)mgkg(-1) in KC-1000. Under PCB-air conditions, residual PCB concentrations were 6.0×10(4)mgkg(-1) and 2.4×10(5)mgkg(-1) in the upper and lower parts for KC-300 and 3.6×10(4)mgkg(-1) and 1.5×10(5)mgkg(-1) in those for KC-1000, respectively, while the rest of the PCBs were infiltrated. On the other hand, when a small amount of PCB oil with the volume far smaller than the total porosity in the column was supplied, the original PCBs were not transported via water permeation. However, lower-chlorinated PCB congeners-e.g., di- or tri-chlorinated biphenyls-preferentially dissolved and were infiltrated from the bottom of the column. These propensities on PCB oil infiltration can be explained in conjunction with the degree of PCB saturation in the sand column.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Water, deionized
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
Supelco
Water, for TOC analysis
Sigma-Aldrich
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Silica, nanopowder, spec. surface area 175-225 m2/g (BET), 99.8% trace metals basis
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O