Merck
OFIRMEV: an old drug becomes new again.
Anesthesia progress (2014-09-06)
Rodney N Nishimoto
RESUMO
This was judged to be the first place winning submission for the American Dental Society of Anesthesiology Student Essay Award. Acetaminophen is an old drug that is now available in an intravenous formulation. Its advantages and disadvantages are reviewed, including its potential role in multimodal postoperative pain therapy.
MATERIAIS
Número do produto
Marca
Descrição do produto
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®