  • Search Results
  • Comparison of superficially porous and fully porous silica supports used for a cyclofructan 6 hydrophilic interaction liquid chromatographic stationary phase.
Comparison of superficially porous and fully porous silica supports used for a cyclofructan 6 hydrophilic interaction liquid chromatographic stationary phase.
Journal of chromatography. A (2014-09-28)
Maressa D Dolzan, Daniel A Spudeit, Zachary S Breitbach, William E Barber, Gustavo A Micke, Daniel W Armstrong
ABSTRACT
A new HILIC stationary phase comprised of native cyclofructan-6 (CF6) bonded to superficially porous silica particles (2.7μm) was developed. Its performance was evaluated and compared to fully porous silica particles with 5μm (commercially available as FRULIC-N) and 3μm diameters. Faster and more efficient chromatography was achieved with the superficially porous particles (SPPs). The columns were also evaluated in the normal phase mode. The peak efficiency, analysis time, resolution, and overall separation capabilities in both HILIC and normal phase modes were compared. The analysis times using the superficially porous based column in HILIC mode were shorter and the theoretical plates/min were higher over the entire range of flow rates studied. The column containing the superficially porous particles demonstrated higher optimum flow rates than the fully porous particle packed columns. At higher flow rates, the advantages of the superficially porous particles was more pronounced in normal phase separations than in HILIC, clearly demonstrating the influence that the mode of chromatography has on band broadening. However, the minimum reduced plate heights (hmin) were typically lower in HILIC than in the normal phase mode. Overall, the superficially porous particle based CF6 column showed clear advantages over the fully porous particle columns, in terms of high throughput and efficient separations of polar compounds in the HILIC mode.
Sigma-Aldrich
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica, nanopowder, spec. surface area 175-225 m2/g (BET), 99.8% trace metals basis
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm