  • Search Results
  • Non-topography-guided PRK combined with CXL for the correction of refractive errors in patients with early stage keratoconus.
Non-topography-guided PRK combined with CXL for the correction of refractive errors in patients with early stage keratoconus.
Journal of refractive surgery (Thorofare, N.J. : 1995) (2014-10-08)
Ali Fadlallah, Ali Dirani, Elias Chelala, Rafic Antonios, George Cherfan, Elias Jarade
ABSTRACT
To evaluate the safety and clinical outcome of combined non-topography-guided photorefractive keratectomy (PRK) and corneal collagen cross-linking (CXL) for the treatment of mild refractive errors in patients with early stage keratoconus. A retrospective, nonrandomized study of patients with early stage keratoconus (stage 1 or 2) who underwent simultaneous non-topography-guided PRK and CXL. All patients had at least 2 years of follow-up. Data were collected preoperatively and postoperatively at the 6-month, 1-year, and 2-year follow-up visit after combined non-topography-guided PRK and CXL. Seventy-nine patients (140 eyes) were included in the study. Combined non-topography-guided PRK and CXL induced a significant improvement in both visual acuity and refraction. Uncorrected distance visual acuity significantly improved from 0.39 ± 0.22 logMAR before combined non-topography-guided PRK and CXL to 0.12 ± 0.14 logMAR at the last follow-up visit (P <.001) and corrected distance visual acuity remained stable (0.035 ± 0.062 logMAR preoperatively vs 0.036 ± 0.058 logMAR postoperatively, P =.79). The mean spherical equivalent decreased from -1.78 ± 1.43 to -0.42 ± 0.60 diopters (D) (P <.001), and the mean cylinder decreased from 1.47 ± 1.10 to 0.83 ± 0.55 D (P <.001). At the last follow-up visit mean keratometry flat was 43.30 ± 1.75 vs 45.62 ± 1.72 D preoperatively (P = .03) and mean keratometry steep was 44.39 ± 3.14 vs 46.53 ± 2.13 D preoperatively (P = .02). Mean central corneal thickness decreased from 501.74 ± 13.11 to 475.93 ± 12.25 µm following combined non-topography-guided PRK and CXL (P < .001). No intraoperative complications occurred. Four eyes developed mild haze that responded well to a short course of topical steroids. No eye developed infectious keratitis. Combined non-topography-guided PRK and CXL is an effective and safe option for correcting mild refractive error and improving visual acuity in patients with early stable keratoconus.
Product Description
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Supelco
Riboflavin (B2), analytical standard
Riboflavin for peak identification, European Pharmacopoeia (EP) Reference Standard