Cytisine versus nicotine for smoking cessation.
The New England journal of medicine (2014-12-18)
Natalie Walker, Colin Howe, Marewa Glover, Hayden McRobbie, Joanne Barnes, Vili Nosa, Varsha Parag, Bruce Bassett, Christopher Bullen
RESUMO
Placebo-controlled trials indicate that cytisine, a partial agonist that binds the nicotinic acetylcholine receptor and is used for smoking cessation, almost doubles the chances of quitting at 6 months. We investigated whether cytisine was at least as effective as nicotine-replacement therapy in helping smokers to quit. We conducted a pragmatic, open-label, noninferiority trial in New Zealand in which 1310 adult daily smokers who were motivated to quit and called the national quitline were randomly assigned in a 1:1 ratio to receive cytisine for 25 days or nicotine-replacement therapy for 8 weeks. Cytisine was provided by mail, free of charge, and nicotine-replacement therapy was provided through vouchers for low-cost patches along with gum or lozenges. Low-intensity, telephone-delivered behavioral support was provided to both groups through the quitline. The primary outcome was self-reported continuous abstinence at 1 month. At 1 month, continuous abstinence from smoking was reported for 40% of participants receiving cytisine (264 of 655) and 31% of participants receiving nicotine-replacement therapy (203 of 655), for a difference of 9.3 percentage points (95% confidence interval, 4.2 to 14.5). The effectiveness of cytisine for continuous abstinence was superior to that of nicotine-replacement therapy at 1 week, 2 months, and 6 months. In a prespecified subgroup analysis of the primary outcome, cytisine was superior to nicotine-replacement therapy among women and noninferior among men. Self-reported adverse events over 6 months occurred more frequently in the cytisine group (288 events among 204 participants) than in the group receiving nicotine-replacement therapy (174 events among 134 participants); adverse events were primarily nausea and vomiting and sleep disorders. When combined with brief behavioral support, cytisine was found to be superior to nicotine-replacement therapy in helping smokers quit smoking, but it was associated with a higher frequency of self-reported adverse events. (Funded by the Health Research Council of New Zealand; Australian New Zealand Clinical Trials Registry number, ACTRN12610000590066.).
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
Sigma-Aldrich
(±)-Nicotine, ≥99% (TLC), liquid
Supelco
S(−)-Nicotine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
(−)-Nicotine, PESTANAL®, analytical standard
Supelco
(−)-Nicotine solution, 1.0 mg/mL, analytical standard, for drug analysis
Sigma-Aldrich
Cytisine, ≥99.0% (HPLC), ≥99%
Sigma-Aldrich
Cytisine, ≥99%, powder
Supelco
(−)-Nicotine solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard