• Início
  • Resultados da busca
  • Pharmacokinetics of cytisine, an α4 β2 nicotinic receptor partial agonist, in healthy smokers following a single dose.
Pharmacokinetics of cytisine, an α4 β2 nicotinic receptor partial agonist, in healthy smokers following a single dose.
Drug testing and analysis (2014-09-19)
Soo Hee Jeong, David Newcombe, Janie Sheridan, Malcolm Tingle
RESUMO
Cytisine, an α4 β2 nicotinic receptor partial agonist, is a plant alkaloid that is commercially extracted for use as a smoking cessation medication. Despite its long history of use, there is very little understanding of the pharmacokinetics of cytisine. To date, no previous studies have reported cytisine concentrations in humans following its use as a smoking cessation agent. A high performance liquid chromatography-ultraviolet (HPLC-UV) method was developed and validated for analysis of Tabex® and nicotine-free oral strips, two commercial products containing cytisine. A sensitive liquid chromatography-mass spectrometry (LC-MS) method was developed and validated for the quantification of cytisine in human plasma and for the detection of cytisine in urine. Single-dose pharmacokinetics of cytisine was studied in healthy smokers. Subjects received a single 3 mg oral dose administration of cytisine. Cytisine was detected in all plasma samples collected after administration, including 15 min post-dose and at 24 h. Cytisine was renally excreted and detected as an unchanged drug. No metabolites were detected in plasma or urine collected in the study. No adverse reactions were reported.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sulfanilamide, ≥98%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥99% (calc. to the dried substance)
Sigma-Aldrich
Cytisine, ≥99.0% (HPLC), ≥99%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Cytisine, ≥99%, powder
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Ultrapure Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Ultrapure Acetonitrile