Merck
Microleakage of inlay ceramic systems luted with self-adhesive resin cements.
The journal of adhesive dentistry (2014-09-30)
Bulent Uludag, Elif Yucedag, Volkan Sahin
RESUMO
To evaluate the microleakage of Cerec 3, IPS e.max Press, and Turkom-Cera inlays cemented with three self-adhesive resin cements. Ninety standardized class III MOD cavities were prepared in intact human mandibular third molars. Ceramic inlays were fabricated according to the manufacturer's instructions and were cemented using three self-adhesive resin cements (RelyX Unicem, Smartcem 2, and SpeedCEM). The specimens were stored in distilled water at 37°C for 24 h and subjected to 1000 thermocycles in water between 5°C and 55°C with a dwell time of 30 s. Subsequently, the specimens were subjected to 100,000 cycles of mechanical loading of 50 N at 1.6 Hz in 37°C water. The specimens were immersed in 0.5% basic fuchsine for 24 h and were sectioned using a low-speed diamond blade. The percentage of dye leakage at the tooth/restoration interface was measured and compared by Kruskal-Wallis tests with Bonferonni correction and Mann-Whitney U-tests at a significance level of p<0.05. Microleakage at the RelyX Unicem interface was lower than that with Smartcem 2 and SpeedCEM resin cements (p<0.05). Microleakage of the Turkom-Cera system was higher than Cerec 3 and IPS e.max Press ceramic inlays (p<0.05). Regardless of the ceramic system and self-adhesive resin cement used, dentin margins were associated with higher microleakage than enamel margins.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, tested according to Ph. Eur.
Supelco
Water, for TOC analysis
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Water, suitable for HPLC
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, deionized
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Supelco
Aluminum oxide, for the determination of hydrocarbons
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O