• Início
  • Resultados da busca
  • Specific recognition of guanines in non-duplex regions of nucleic acids with potassium tungstate and hydrogen peroxide.
Specific recognition of guanines in non-duplex regions of nucleic acids with potassium tungstate and hydrogen peroxide.
Nucleic acids research (2014-10-31)
Wuxiang Mao, Xiaowei Xu, Huan He, Rong Huang, Xi Chen, Heng Xiao, Zhenduo Yu, Yi Liu, Xiang Zhou
RESUMO
Structural features of nucleic acids have become an integral part of current biomedical research. Highly selective and readily performed methods with little toxicity that target guanosines in non-duplex nucleic acids are needed, which led us to search for an effective agent for guanosine sequencing. Treatment of DNA or RNA with potassium tungstate and hydrogen peroxide produced damaged guanosines in DNA or RNA sequences. The damaged guanosines in non-duplex DNA could be cleaved by hot piperidine. Similarly, damaged guanosines in non-duplex RNA could be cleaved by aniline acetate. We could identify structural features of nucleic acid using this strategy instead of dimethyl sulphate and Ribonuclease T1.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Guanine, 98%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Supelco
Guanine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Guanine, BioUltra