• Início
  • Resultados da busca
  • Co-culture of endothelial cells and patterned smooth muscle cells on titanium: construction with high density of endothelial cells and low density of smooth muscle cells.
Co-culture of endothelial cells and patterned smooth muscle cells on titanium: construction with high density of endothelial cells and low density of smooth muscle cells.
Biochemical and biophysical research communications (2014-12-03)
Jingan Li, Kun Zhang, Juejue Wu, Yuzhen Liao, Ping Yang, Nan Huang
RESUMO
Endothelialization has been considered a promising method to improve the biocompatibility of vascular implanted biomaterials. However, little is known about the anti-coagulation, anti-inflammatory, anti-atherosclerosis and anti-shedding property of the attached endothelial cells (ECs) and the relationship with their bio-environment and material-environment, which are both important evaluations to the cardiovascular biomaterials designed for tissue engineering applications and in vivo implantation. In this in vitro study, a novel co-culture model was built, where vascular smooth muscle cells (SMCs) were cultured on the hyaluronic acid (HA) micro-strip patterned titanium (Ti) surface on a low density to biomimetic the EC pericyte environment. Subsequently, the EC number and its functional factor, including nitric oxide (NO), prostacyclin (PGI2), tissue factor pathway inhibitor (TFPI), thrombomodulin (TM), and the inflammatory induced factor, endothelial leukocyte adhesion molecule-1 (E-selectin) were quantified, respectively. The anti-shedding property was also assessed by the blood flow shear stress (BFSS) acting. The results showed that the novel co-culture model possessed better EC coverage, functional factor release and anti-shedding functions than the control.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, powder, <45 μm avg. part. size, 99.98% trace metals basis
Sigma-Aldrich
Titanium, powder, −100 mesh, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, sponge, 3-19 mm, 99.5% trace metals basis
Sigma-Aldrich
Titanium, nanoparticles, dispersion, <100 nm particle size, in mineral oil, 98.5% trace metals basis
Sigma-Aldrich
Titanium, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.995% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, crystalline, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 2.0 mm, 99.99% trace metals basis
Titanium, rod, 100mm, diameter 10mm, annealed, 99.6+%
Titanium, tube, 200mm, outside diameter 0.51mm, inside diameter 0.35mm, wall thickness 0.08mm, annealed, 99.6+%
Sigma-Aldrich
Titanium, wire, diam. 0.127 mm, 99.99% trace metals basis
Titanium, rod, 100mm, diameter 2mm, annealed, 99.6+%
Titanium, rod, 500mm, diameter 2mm, as drawn, 99.99+%
Titanium, wire reel, 1m, diameter 0.25mm, as drawn, 99.6+%
Titanium, wire reel, 1m, diameter 0.5mm, as drawn, 99.6+%
Titanium, tube, 500mm, outside diameter 0.51mm, inside diameter 0.35mm, wall thickness 0.08mm, annealed, 99.6+%
Titanium, tube, 500mm, outside diameter 0.81mm, inside diameter 0.61mm, wall thickness 0.1mm, as drawn, 99.6+%
Titanium, tube, 500mm, outside diameter 1.05mm, inside diameter 0.75mm, wall thickness 0.15mm, annealed, 99.6+%
Titanium, tube, 500mm, outside diameter 1.6mm, inside diameter 1.2mm, wall thickness 0.2mm, hard, 99.6+%