  • Search Results
  • Low- and high-grade bladder cancer appraisal via serum-based proteomics approach.
Low- and high-grade bladder cancer appraisal via serum-based proteomics approach.
Clinica chimica acta; international journal of clinical chemistry (2014-05-31)
Navneeta Bansal, Ashish Gupta, Satya Narain Sankhwar, Abbas Ali Mahdi
ABSTRACT
To address the shortcomings of urine cytology and cystoscopy for screening and grading of urinary bladder cancer (BC) we applied a serum-based proteomics approach as a surrogate tactic for rapid BC probing. This study was performed on 90 sera samples comprising of low-grade (LG, n=33) and high-grade (HG, n=32) BC, and healthy controls (HC, n=25). Two-dimensional gel electrophoresis (2DE) tactic was executed to describe serum proteome. MALDI-TOF-MS (MS) was used to identify the characteristics of aberrantly expressed proteins in 2DE and validated using Western blot (WB) and ELISA approach. Receiver operating characteristics (ROC) curve analysis was also performed to determine the clinical usefulness of these proteins to discriminate among LG, HG and HC cohorts. This comprehensive approach of 2DE, MS, WB and ELISA reveals five differentially expressed proteins. Among them two biomarkers (S100A8 and S100A9) were able to accurately (ROC, 0.946) distinguish 81% of BC (LG+HG) cases compared to HC with highest sensitivity and specificity. With a comparable tactic, two biomarkers (S100A8 and S100A4) were able to precisely (ROC, 0.941) discriminate 92% of LG cases from HG with utmost sensitivity and specificity. Serum proteomics probing appears to be an encouraging and least-invasive tactic for screening and grading of BC.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Ultrapure Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC