Merck
  • Início
  • Resultados da busca
  • Development of an in vitro cytochrome P450 cocktail inhibition assay for assessing the inhibition risk of drugs of abuse.
Development of an in vitro cytochrome P450 cocktail inhibition assay for assessing the inhibition risk of drugs of abuse.
Toxicology letters (2014-08-12)
Julia Dinger, Markus R Meyer, Hans H Maurer
RESUMO
Drugs of abuse are not tested for cytochrome P450 (CYP) inhibition potential before distribution. Therefore, a cocktail assay should be developed for testing the inhibition potential for all relevant CYPs. The following CYP test substrates and selective inhibitors were incubated in pooled human liver microsomes: phenacetin (alpha-naphthoflavone for CYP1A2), coumarin (tranylcypromine, CYP2A6), bupropion (sertraline, CYP2B6), amodiaquine (trimethoprim, CYP2C8), diclofenac (sulfaphenazole, CYP2C9), omeprazole (fluconazole, CYP2C19), dextromethorphan (quinidine, CYP2D6), chlorzoxazone (clomethiazole, CYP2E1), testosterone (verapamil, CYP3A). Samples were analyzed after protein precipitation using a Thermo Fisher Q-Exactive LC-high-resolution-MS/MS. The IC50 values were calculated by plotting the concentration of the formed metabolite, relative to the control sample, over the logarithm of the inhibitor concentration. They were determined either for single substrate or the cocktail incubation. Unfortunately, the cocktail assay had to be split because of interferences during incubation caused by substrates or metabolites, but the mixture of both incubates could be analyzed in one analytical run. The IC50 values determined in the single substrate or both cocktail incubations were comparable among themselves and with published data. In conclusion, the new inhibition cocktail assay was reproducible and applicable for testing the inhibition potential of drugs of abuse as exemplified for 2,5-dimethoxy-4-iodo-amfetamine (DOI).
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Sulfaphenazole, ≥98%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Quinidine, anhydrous
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Quinidine, crystallized, ≥98.0% (dried material, NT)
Sigma-Aldrich
Chlorzoxazone
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Ultrapure Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
trans-2-Phenylcyclopropylamine hydrochloride, 97%
Fluconazole for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Trimethoprim for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Sulfaphenazole, ≥98% (HPLC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%