Merck
  • Início
  • Resultados da busca
  • Ultrastructure of human tracheal smooth muscle from subjects with asthma and nonasthmatic subjects. Standardized methods for comparison.
Ultrastructure of human tracheal smooth muscle from subjects with asthma and nonasthmatic subjects. Standardized methods for comparison.
American journal of respiratory cell and molecular biology (2014-07-24)
Harley T Syyong, Chris D Pascoe, Jenny Zhang, Bryna A Arsenault, Dennis Solomon, W Mark Elliott, Tillie L Hackett, David C Walker, Peter D Paré, Chun Y Seow
RESUMO
A characteristic feature of asthma is exaggerated airway narrowing, termed airway hyper-responsiveness (AHR) due to contraction of airway smooth muscle (ASM). Although smooth muscle (SM)-specific asthma susceptibility genes have been identified, it is not known whether asthmatic ASM is phenotypically different from nonasthmatic ASM in terms of subcellular structure or mechanical function. The present study is the first to systematically quantify, using electron microscopy, the ultrastructure of tracheal SM from subjects with asthma and nonasthmatic subjects. Methodological details concerning tissue sample preparation, ultrastructural quantification, and normalization of isometric force by appropriate morphometric parameters are described. We reasoned that genetic and/or acquired differences in the ultrastructure of asthmatic ASM could be associated with functional changes. We recently reported that asthmatic ASM is better able to maintain and recover active force generation after length oscillations simulating deep inspirations. The present study was designed to seek structural evidence to account for this observation. Contrary to our hypotheses, no significant qualitative or quantitative differences were found in the subcellular structure of asthmatic versus nonasthmatic tracheal SM. Specifically, there were no differences in average SM cell cross-sectional area; fraction of the cell area occupied by nonfilamentous area; amounts of mitochondria, dense bodies, and dense plaques; myosin and actin filament densities; basal lamina thickness; and the number of microtubules. These results indicate that functional differences in ASM do not necessarily translate into observable structural changes.
MATERIAIS
Número do produto
Marca
Descrição do produto
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®