• Início
  • Resultados da busca
  • Cadmium and proliferation in human uterine leiomyoma cells: evidence of a role for EGFR/MAPK pathways but not classical estrogen receptor pathways.
Cadmium and proliferation in human uterine leiomyoma cells: evidence of a role for EGFR/MAPK pathways but not classical estrogen receptor pathways.
Environmental health perspectives (2014-10-25)
Xiaohua Gao, Linda Yu, Alicia B Moore, Grace E Kissling, Michael P Waalkes, Darlene Dixon
RESUMO
It has been proposed that cadmium (Cd) is an environmental "metalloestrogen" and that its action is mediated via the estrogen receptor (ER). Cd mimics the effects of estrogen in the rat uterus, and blood Cd concentrations positively correlate with ER levels in uteri of women with fibroids. In the present study we explored whether Cd could stimulate proliferation of estrogen-responsive human uterine leiomyoma (ht-UtLM) cells and uterine smooth muscle cells (ht-UtSMCs) through classical interactions with ERα and ERβ, or by nongenomic mechanisms. We used estrogen response element (ERE) reporters, phosphorylated receptor tyrosine kinase arrays, Western blot analysis, estrogen binding, and cell proliferation assays to evaluate the effects of Cd on ht-UtLM cells and ht-UtSMCs. Cd stimulated growth of both cell types at lower concentrations and inhibited growth at higher concentrations (≥ 50 μM). Cd did not significantly bind to ERα or ERβ, nor did it show transactivation in both cell types transiently transfected with ERE reporter genes. However, in both cells types, Cd (0.1 μM and 10 μM) activated p44/42 MAPK (ERK1/2), and a MAPK inhibitor (PD98059) abrogated Cd-induced cell proliferation. Cd in ht-UtLM cells, but not in ht-UtSMCs, activated the growth factor receptors EGFR, HGFR, and VEGF-R1 upstream of MAPK. Additional studies in ht-UtLM cells showed that AG1478, an EGFR inhibitor, abolished Cd-induced phosphorylation of EGFR and MAPK. Our results show that low concentrations of Cd stimulated cell proliferation in estrogen-responsive uterine cells by nongenomic activation of MAPK, but not through classical ER-mediated pathways.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Guanine, 98%
Sigma-Aldrich
Cadmium chloride, 99.99% trace metals basis
Supelco
Guanine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cadmium chloride, technical grade
Sigma-Aldrich
PD 98,059, solid
Sigma-Aldrich
Tyrphostin AG 1478, ≥98%
Tyrosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Guanine, BioUltra
Sigma-Aldrich
DL-Tyrosine, 99%