Merck
  • Início
  • Resultados da busca
  • Identification of new MRP4 inhibitors from a library of FDA approved drugs using a high-throughput bioluminescence screen.
Identification of new MRP4 inhibitors from a library of FDA approved drugs using a high-throughput bioluminescence screen.
Biochemical pharmacology (2014-12-03)
Leanna Cheung, Denise M T Yu, Zillan Neiron, Tim W Failes, Greg M Arndt, Jamie I Fletcher
RESUMO
Multidrug resistance protein 4 (MRP4) effluxes a wide variety of drugs and endogenous signaling molecules from cells and has been proposed as an attractive therapeutic target in several solid tumors, including neuroblastoma and colorectal cancer. MRP4 also regulates the pharmacokinetics of its drug substrates and its absence can increase their tissue penetration. We observed that MRP4 can efflux the bioluminescence substrate d-luciferin, and exploited this phenomenon to develop a robust, high throughput, live cell-based bioluminescent screen to identify new MRP4 inhibitors. We applied this screen to a combined library of 3600 compounds, all of which were either FDA-approved drugs or bioactive compounds with defined mechanisms of action. From the primary screen, 36 compounds effectively inhibited MRP4 (>4-fold increase in bioluminescence), with inhibitors of receptor tyrosine kinases and phosphodiesterases highly over-represented. Selected compounds were tested for their ability to sensitize MRP4-overexpressing cell lines to the MRP4 substrate drugs 6-mercaptopurine and SN-38, with sensitization up to 6.5-fold with the ryanodine receptor antagonist dantrolene. These newly identified MRP4 inhibitors are readily available and are either established drugs or well-characterized bioactive compounds. As such, they should be immediately useful as investigative tools, and suitable for testing both in vitro and in vivo.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Supelco
Glafenine hydrochloride, analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dantrolene sodium salt
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O