  • Search Results
  • Differential response of glial fibrillary acidic protein-positive astrocytes in the rat prefrontal cortex following ethanol self-administration.
Differential response of glial fibrillary acidic protein-positive astrocytes in the rat prefrontal cortex following ethanol self-administration.
Alcoholism, clinical and experimental research (2015-04-04)
Cecilia Bull, Wahab A Syed, Sabrina C Minter, M Scott Bowers
ABSTRACT
Prefrontal cortex (PFC) dysfunction is believed to contribute to the transition from controlled substance use to abuse. Because astrocytes have been suggested to play a key role in the development and maintenance of drug-seeking behaviors, we sought to determine whether PFC astrocytes are affected by ethanol (EtOH) self-administration. EtOH consumption was modeled in rats by 3 self-administration paradigms where EtOH was made concurrently available with water in the home cage either continuously (CEA) or intermittently (IEA). In the third paradigm, EtOH was only available in the operant chamber (OEA). To avoid the potential confound of acute EtOH effects, all rats were sacrificed after either 24-hour or 3-week abstinence. In all groups, the effect of EtOH consumption on PFC astrocytes was measured using unbiased stereological counting of cells expressing the astrocyte marker glial fibrillary acidic protein (GFAP). GFAP immunoreactivity commonly changes in response to pharmacological insult or injury. GFAP-positive astrocyte number increased in the prelimbic and anterior cingulate cortex regions of the PFC after IEA. No change was found in the infralimbic or orbitofrontal cortex after IEA. After 3-week abstinence, there was a reduction of astrocytes in the prelimbic and orbitofrontal cortex of the CEA cohort as well as a reduction in the orbitofrontal cortex of the OEA cohort. These findings demonstrate that discrete PFC subregions contain GFAP-positive astrocyte populations that respond differentially to distinct EtOH consumption paradigms. A better understanding of how specific astrocyte populations uniquely adapt to EtOH consumption could provide insight for targeted therapeutic interventions.
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone and 0.5% 4-methyl-2-pentanone, S15, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Supelco
o-Xylene, analytical standard
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
2-Chloroethyl acrylate, contains >100 ppm MEHQ as inhibitor, 97%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required
Sigma-Aldrich
Ethyl alcohol, Pure, 160 proof, Excise Tax-free, Permit for use required
Supelco
o-Xylene, suitable for HPLC, 98%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 1% cyclohexane, A15 CYCLO1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
3-(2-Hydroxyethyl)indole, 97%