  • Search Results
  • Reduction of tendon adhesions following administration of Adaprev, a hypertonic solution of mannose-6-phosphate: mechanism of action studies.
Reduction of tendon adhesions following administration of Adaprev, a hypertonic solution of mannose-6-phosphate: mechanism of action studies.
PloS one (2014-11-11)
Jason K F Wong, Anthony D Metcalfe, Richard Wong, Jim Bush, Chris Platt, Arnaud Garcon, Nick Goldspink, Duncan A McGrouther, Mark W J Ferguson
ABSTRACT
Repaired tendons may be complicated by progressive fibrosis, causing adhesion formation or tendon softening leading to tendon rupture and subsequent reduced range of motion. There are few therapies available which improve the gliding of damaged tendons in the hand. We investigate the role of Mannose 6-phosphate (M6P) in a 600 mM hypertonic solution (Adaprev) on tendon adhesion formation in vivo using a mouse model of severed tendon in conjunction with analysis of collagen synthesis, cellular proliferation and receptors involved in TGF beta signalling. Cytotoxicity was assessed by measuring tissue residency, mechanical strength and cell viability of tendons after treatment with Adaprev. To elicit potential modes of action, in vitro and ex vivo studies were performed investigating phosphorylation of p38, cell migration and proliferation. Adaprev treatment significantly (p<0.05) reduced the development of adhesions and improved collagen organisation without reducing overall collagen synthesis following tendon injury in vivo. The bioavailability of Adaprev saw a 40% reduction at the site of administration over 45 minutes and tendon fibroblasts tolerated up to 120 minutes of exposure without significant loss of cell viability or tensile strength. These favourable effects were independent of CI-MPR and TGF-β signalling and possibly highlight a novel mechanism of action related to cellular stress demonstrated by phosphorylation of p38. The effect of treatment reduced tendon fibroblast migration and transiently halted tendon fibroblast proliferation in vitro and ex vivo. Our studies demonstrate that the primary mode of action for Adaprev is potentially via a physical, non-chemical, hyperosmotic effect.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Eosin Y disodium salt, certified by the Biological Stain Commission
Sigma-Aldrich
Eosin Y disodium salt, Dye content ≥85 %
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Eosin Y
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Eosin Y solution, 5 wt. % in H2O
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Calcein-AM, BioReagent, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Supelco
Methanol, analytical standard
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 20-40 mesh, reagent grade, 97%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)