• Início
  • Resultados da busca
  • Differences in IGF axis-related proteins in amniotic fluid of trisomy 21 and trisomy 18 using a multiple reaction monitoring approach.
Differences in IGF axis-related proteins in amniotic fluid of trisomy 21 and trisomy 18 using a multiple reaction monitoring approach.
Prenatal diagnosis (2014-07-02)
Guangrui Lai, Yuanyuan Zhang, Leitong Wang, Bijun Zhang, Gaoyan Zheng, Xuanyi Wang, Xiaoqiu Zhao, Haiming Gao, Yanyan Zhao
RESUMO
Trisomy 21 and trisomy 18 are the two most common chromosomal anomalies in live births. To find new biomarkers for aneuploidies and pathogenesis of fetal malformations, we measured insulin-like growth factor (IGF) axis-related proteins in amniotic fluid (AF) of pregnant women carrying trisomies 21 or 18 affected fetuses using multiple reaction monitoring (MRM) approach. Eighty-five AF samples from pregnant women carrying either trisomy 21, trisomy 18, or normal fetuses were collected. IGF axis-related proteins in AF after serial treatments were quantitated with MRM method. The differential protein levels were also confirmed by western blot in AF without any treatment. The IGF type I receptor and pregnancy-associated plasma protein-A in AF of trisomy 21 (1.35 ± 0.32 and 13.36 ± 3.64 µg/mg protein) and trisomy 18 (1.39 ± 0.40 and 12.80 ± 1.84 µg/mg protein) were decreased versus normal controls (2.16 ± 0.59 and 23.77 ± 6.18 µg/mg protein). IGF binding protein 5 was reduced in trisomy 18 (1.47 ± 0.33 vs 2.36 ± 0.77 µg/mg protein). These alterations were confirmed by western blot. The other proteins showed no significant difference between the three groups. Our data suggested that MRM can provide a powerful platform for the identification of biomarkers in AF that have crucial developmental effects in the aneuploid fetus.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Ammonium bicarbonate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%