• Início
  • Resultados da busca
  • Pharmacokinetics and safety of rifabutin in young HIV-infected children receiving rifabutin and lopinavir/ritonavir.
Pharmacokinetics and safety of rifabutin in young HIV-infected children receiving rifabutin and lopinavir/ritonavir.
The Journal of antimicrobial chemotherapy (2014-10-05)
Harry Moultrie, Helen McIlleron, Shobna Sawry, Tracy Kellermann, Lubbe Wiesner, Gurpreet Kindra, Hermien Gous, Annelies Van Rie
RESUMO
Co-treatment of HIV and TB in young children is complicated by limited treatment options and complex drug-drug interactions. Rifabutin is an alternative to rifampicin for adults receiving a ritonavir-boosted PI. We aimed to evaluate the short-term safety and pharmacokinetics of rifabutin when given with lopinavir/ritonavir in children. We conducted an open-label study of rifabutin dosed at 5 mg/kg three times a week in HIV-infected children≤5 years of age receiving lopinavir/ritonavir. Intensive steady-state pharmacokinetic sampling was conducted after six doses. The Division of AIDS 2004, clarification 2009, table for grading severity of adverse events was used to classify drug toxicities. The study was registered with ClinicalTrials.gov, number NCT01259219. Six children completed the study prior to closure by institutional review boards. The median (range) AUC0-48 of rifabutin was 6.91 (3.52-8.67) μg · h/mL, the median (range) Cmax of rifabutin was 0.39 (0.19-0.46) μg/mL, the median (range) AUC0-48 of 25-O-desacetyl rifabutin was 5.73 (2.85-9.13) μg · h/mL and the median (range) Cmax of 25-O-desacetyl rifabutin was 0.17 (0.08-0.32) μg/mL. The neutrophil count declined in all children; two children experienced grade 4 neutropenia, which resolved rapidly without complications. There was strong correlation between AUC0-48 measures and neutrophil counts. Rifabutin dosed at 5 mg/kg three times per week resulted in lower AUC0-48, AUC0-24 and Cmax values for rifabutin and 25-O-desacetyl rifabutin compared with adults receiving 150 mg of rifabutin daily, the current recommended dose. We observed high rates of severe transient neutropenia, possibly due to immaturity of CYP3A4 in young children. It remains unclear whether a safe and effective rifabutin dose exists for treatment of TB in children receiving lopinavir/ritonavir.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Ultrapure Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, for DNA synthesis