  • Search Results
  • Irradiation with X-rays phase-advances the molecular clockwork in liver, adrenal gland and pancreas.
Irradiation with X-rays phase-advances the molecular clockwork in liver, adrenal gland and pancreas.
Chronobiology international (2014-08-21)
Mareike Hildegard Müller, Franz Rödel, Udo Rüb, Horst-Werner Korf
ABSTRACT
The circadian clock of man and mammals shows a hierarchic organization. The master clock, located in the suprachiasmatic nuclei (SCN), controls peripheral oscillators distributed throughout the body. Rhythm generation depends on molecular clockworks based on transcriptional/translational interaction of clock genes. Numerous studies have shown that the clockwork in peripheral oscillators is capable to maintain circadian rhythms for several cycles in vitro, i.e. in the absence of signals from the SCN. The aim of the present study is to analyze the effects of irradiation with X-rays on the clockwork of liver, adrenal and pancreas. To this end organotypic slice cultures of liver (OLSC) and organotypic explant cultures of adrenal glands (OAEC) and pancreas (OPEC) were prepared from transgenic mPer2(luc) mice which express luciferase under the control of the promoter of an important clock gene, Per2, and allow to study the dynamics of the molecular clockwork by bioluminometry. The preparations were cultured in a membrane-based liquid-air interface culturing system and irradiated with X-rays at doses of 10 Gy and 50 Gy or left untreated. Bioluminometric real-time recordings show a stable oscillation of all OLSC, OAEC and OPEC for up to 12 days in vitro. Oscillations persist after irradiation with X-rays. However, a dose of 50 Gy caused a phase advance in the rhythm of the OLSC by 5 h, in the OPEC by 7 h and in the OAEC by 6 h. Our study shows that X-rays affect the molecular clockwork in liver, pancreas and adrenal leading to phase advances. Our results confirm and extend previous studies showing a phase-advancing effect of X-rays at the level of the whole animal and single cells.
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)