• Início
  • Resultados da busca
  • Quantification of thiazolidine-4-carboxylic acid in toxicant-exposed cells by isotope-dilution liquid chromatography-mass spectrometry reveals an intrinsic antagonistic response to oxidative stress-induced toxicity.
Quantification of thiazolidine-4-carboxylic acid in toxicant-exposed cells by isotope-dilution liquid chromatography-mass spectrometry reveals an intrinsic antagonistic response to oxidative stress-induced toxicity.
Chemical research in toxicology (2014-10-18)
Jingjing Liu, Wan Chan
RESUMO
Carcinogenic formaldehyde is produced by endogenous protein oxidation and various exogenous sources. With formaldehyde being both ubiquitous in the ambient environment and one of the most common reactive carbonyls produced from endogenous metabolism, quantifying formaldehyde exposure is an essential step in risk assessments. We present in this study an approach to assess the risk of exposure to oxidative stress by quantifying thiazolidine-4-carboxylic acid (TA), a cysteine-conjugated metabolite of formaldehyde in toxicant-exposed Escherichia coli. The method entails TA derivatization with ethyl chloroformate, addition of isotope-labeled TA derivatives as internal standards, solid-phase extraction of the derivatives, and quantification by liquid chromatography-mass spectrometry (LC-MS). After validating for accuracy and precision, the developed method was used to detect TA in oxidizing agent-exposed E. coli samples. Dose-dependent TA formation was observed in E. coli exposed to hydroxyl radical mediators Fe(2+)-EDTA, H2O2, and NaOCl, indicating the potential use of TA as a biomarker of exposure to oxidative stress and disease risk.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
SAFC
L-Cysteine
Sigma-Aldrich
Ethyl chloroformate, 97%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
L-Cysteine, certified reference material, TraceCERT®