  • Search Results
  • A sensitive sequential 'on/off' SERS assay for heparin with wider detection window and higher reliability based on the reversed surface charge changes of functionalized Au@Ag nanoparticles.
A sensitive sequential 'on/off' SERS assay for heparin with wider detection window and higher reliability based on the reversed surface charge changes of functionalized Au@Ag nanoparticles.
Biosensors & bioelectronics (2014-12-03)
Yi Zeng, Jin-Ju Pei, Li-Hua Wang, Ai-Guo Shen, Ji-Ming Hu
ABSTRACT
A sequential 'on/off' dual mode SERS assay platform for heparin with wider detection window and higher reliability is constructed based on electrostatic forces, in which the highly protonated chitosan encapsulated p-Mercaptobenzoic acid coated Au@Ag core-shell nanoparticles undergo sequential aggregation/segregation upon the additive of heparin with a limit of detection of 43.74ng/mL (5.69U/mL) and a continuous concentration range of 50-800ng/mL (6.5-104U/mL), which are lower in sensitivity and wider in detection window than the most reported assay for heparin. Remarkably, the latter declined window over a range of 350-800ng/mL in contrast, which has not reported before, is extremely important in reliable and practical assay of heparin.
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, hydrogen-storage grade, 98%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, 90% NaBH4 basis, pellets, diam. 8 mm, Cobalt (Doped)
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
4-Mercaptobenzoic acid, 99%
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium borohydride solution, 0.5 M in 2-methoxyethyl ether
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG