Investigation of a nanosuspension stabilized by Soluplus® to improve bioavailability.
International journal of pharmaceutics (2014-12-03)
Hua Yang, Fei Teng, Puxiu Wang, Bin Tian, Xia Lin, Xi Hu, Ling Zhang, Keru Zhang, Yu Zhang, Xing Tang
RESUMO
The purpose of this work was to explore the feasibility of using Soluplus(®) in preparing a fenofibrate (FBT) nanosuspension adopting wet media milling technology. HPMC and Soluplus(®) were used as stabilizers to prepare FBT/HPMC nanosuspension (F1) and FBT/Soluplus(®) nanosuspension (F2), respectively. The nanosuspensions were subjected to evaluations involving particle size, dissolution, preliminary stability and pharmacokinetic behavior. A marked reduction in particle size was achieved by nanosuspensions (from 17.55 μm to 642 nm (F1) and 344 nm (F2)). The nanosuspensions displayed almost complete dissolution while percentages of 30% and 13% were obtained by physical mixtures and coarse FBT separately. Soluplus(®) could stabilize the nanosuspension more effectively due to a weaker Ostwald ripening effect resulting from a slower diffusion of micelles formed by Soluplus(®) entrapping dissolved FBT than FBT exposed to pure water directly. In the in vivo evaluation, larger AUC0-72h and Cmax, and shorter Tmax were obtained by the nanosuspensions. Significant differences were observed between the physical mixtures. The phenomenon of double peaks was present in this study. The major factor may be the multiple absorption sites of FBT. The current work indicated that Soluplus(®) is well suited for preparation of a nanosuspension with good stability and improved dissolution and bioavailability.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Ultrapure Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, for DNA synthesis