  • Search Results
  • Comparison of the adipogenesis in intramuscular and subcutaneous adipocytes from Bamei and Landrace pigs.
Comparison of the adipogenesis in intramuscular and subcutaneous adipocytes from Bamei and Landrace pigs.
Biochemistry and cell biology = Biochimie et biologie cellulaire (2014-06-20)
Guo Hua Zhang, Jian Xiong Lu, Yan Chen, Yong Qing Zhao, Peng Hui Guo, Ju Tian Yang, Rong Xin Zang
ABSTRACT
Fat deposition is a complex process involving proliferation, differentiation, and lipogenesis of adipocytes. Bamei and Landrace are considered to represent fat- and lean-type pig breeds. Subcutaneous (SC) and intramuscular (IM) pre-adipocytes were cultured to compare the proliferation and lipogenesis in these breeds. The differentiated adipocytes were exposed to glucose or insulin to evaluate their effects on lipogenesis and lipogenic gene expression. Pre-adipocytes proliferated dramatically faster in SC vs. IM cells, and in Bamei vs. Landrace breeds. Lipogenesis and lipogenic gene expression had a greater increase in Bamei than in Landrace, and in SC vs. IM in the process of differentiation. Glucose markedly promoted lipogenesis and lipogenic gene expression in differentiated adipocytes. The stimulation of high-glucose levels on lipogenesis and ChREBP and lipogenic gene expression was higher in SC than IM adipocytes, and in Bamei vs. Landrace. Insulin largely increased SREBP-1c expression, however it modestly stimulated lipogenesis and lipogenic gene expression, and there was no difference between cell populationsor between breeds. These data demonstrated that regional and varietal differences obviously existed in the development of porcine adipocytes. The proliferation and differentiation capacity of pre-adipocytes, and the adipocyte lipogenesis stimulated by glucose, are stronger in Bamei than Landrace, and in SC vs. IM adipocytes independent of breed.
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99%, BioUltra
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard