Merck
  • Início
  • Resultados da busca
  • Long-term trends of PBDEs, triclosan, and triclocarban in biosolids from a wastewater treatment plant in the Mid-Atlantic region of the US.
Long-term trends of PBDEs, triclosan, and triclocarban in biosolids from a wastewater treatment plant in the Mid-Atlantic region of the US.
Journal of hazardous materials (2014-10-06)
Natasha A Andrade, Nuria Lozano, Laura L McConnell, Alba Torrents, Clifford P Rice, Mark Ramirez
RESUMO
In the US, land application of biosolids has been utilized in government-regulated programs to recycle valuable nutrients and organic carbon that would otherwise be incinerated or buried in landfills. While many benefits have been reported, there are concerns that these practices represent a source of organic micropollutants to the environment. In this study, biosolids samples from a wastewater treatment plant in the Mid-Atlantic region of the US were collected approximately every 2 months over a 7-year period and analyzed for brominated diphenyl ethers (BDE-47, BDE-99, and BDE-209), triclosan, and triclocarban. During the collection period of 2005-2011, concentrations of the brominated diphenyl ethers BDE-47+BDE-99 decreased by 42%, triclocarban decreased by 47%, but BDE-209 and triclosan remained fairly constant. Observed reductions in contaminant concentrations could not be explained by different seasons or by volumetric changes of wastewaters arriving at the treatment plant and instead may be the result of the recent phaseout of BDE-47 and BDE-99 as well as potential reductions in the use of triclocarban.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Supelco
Dichloromethane, analytical standard
Supelco
Aluminum oxide, for the determination of hydrocarbons
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane solution, certified reference material, 5000 μg/mL in methanol
Supelco
Dichloromethane solution, certified reference material, 200 μg/mL in methanol
Supelco
Dichloromethane, analytical standard, stabilized
Methylene chloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Aluminum oxide, activated, basic, Brockmann I
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminum oxide, powder, primarily α phase, ≤10 μm avg. part. size, 99.5% trace metals basis
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, pore size 58 Å, ~150 mesh
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer