  • Search Results
  • A shotgun approach for the identification of platinum-protein complexes.
A shotgun approach for the identification of platinum-protein complexes.
Analytical and bioanalytical chemistry (2015-01-27)
Irene Moraleja, Estefanía Moreno-Gordaliza, Diego Esteban-Fernández, M Luz Mena, Michael W Linscheid, M Milagros Gómez-Gómez
ABSTRACT
A shotgun approach including peptide-based OFFGEL-isoelectric focusing (IEF) fractionation has been developed with the aim of improving the identification of platinum-binding proteins in biological samples. The method is based on a filter-aided sample preparation (FASP) tryptic digestion under denaturing and reducing conditions of cisplatin-, oxaliplatin-, and carboplatin-protein complexes, followed by OFFGEL-IEF separation of the peptides. Any risk of platinum loss is minimized throughout the procedure due to the removal of the reagents used after each stage of the FASP method and the absence of thiol-based reagents in the focusing buffer employed in the IEF separation. The platinum-peptide complexes stability after the FASP digestion and the IEF separation was confirmed by size exclusion chromatography-inductively coupled plasma-mass spectrometry (SEC-ICP-MS). The suitability of peptide-based OFFGEL-IEF fractionation for reducing the sample complexity for further nano-liquid chromatography-electrospray ionization-tandem mass spectrometry (nLC-ESI-MS/MS) analysis has been demonstrated, allowing the detection of platinum-containing peptides, with significantly lower abundance and ionization efficiency than unmodified peptides. nLC-MS/MS analysis of selected OFFGEL-IEF fractions from tryptic digests with different complexity degrees: standard human serum albumin (HSA), a mixture of five proteins (albumin, transferrin, carbonic anhydrase, myoglobin, and cytochrome-c) and human blood serum allowed the identification of several platinum-peptides from cisplatin-HSA. Cisplatin-binding sites in HSA were elucidated from the MS/MS spectra and assessed considering the protein three-dimensional structure. Most of the potential superficial binding sites available on HSA were identified for all the samples, including a biologically relevant cisplatin-cross-link of two protein domains, demonstrating the capabilities of the methodology.
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis