  • Search Results
  • Induction of DNA damage and G2 cell cycle arrest by diepoxybutane through the activation of the Chk1-dependent pathway in mouse germ cells.
Induction of DNA damage and G2 cell cycle arrest by diepoxybutane through the activation of the Chk1-dependent pathway in mouse germ cells.
Chemical research in toxicology (2015-01-31)
Jianyun Dong, Zhi Wang, Peng Zou, Guowei Zhang, Xiaomei Dong, Xi Ling, Xi Zhang, Jinyi Liu, Dongqing Ye, Jia Cao, Lin Ao
ABSTRACT
1,2:3,4-Diepoxybutane (DEB) is a major carcinogenic metabolite of 1,3-butadiene (BD), which has been shown to cause DNA strand breaks in cells through its potential genotoxicity. The adverse effect of DEB on male reproductive cells in response to DNA damage has not been thoroughly studied, and the related mechanism is yet to be elucidated. Using mouse spermatocyte-derived GC-2 cells, we demonstrated in the present study that DEB caused the proliferation inhibition and marked cell cycle arrest at the G2 phase but not apoptosis. DEB also induced DNA damage as evidenced by γ-H2AX expression, the comet assay, and the cytokinesis-block micronucleus assay. Meanwhile, DEB triggered the Chk1/Cdc25c/Cdc2 signal pathway, which could be abated in the presence of UCN-01 or Chk1 siRNA. GC-2 cells exposed to DEB experienced ROS generation and pretreatment of N-acetyl-l-cysteine, partly attenuated DEB-induced DNA damage, and G2 arrest. Furthermore, measurement of testicular cells showed an increased proportion of tetraploid cells in mice administrated with DEB, alongside the enhanced expression of p-Chk1. Also, the defective reproductive phenotypes, including reduced sperm motility, increased sperm malformation, and histological abnormality of testes, were observed. In conclusion, these results suggest DEB induces DNA damage and G2 cell cycle arrest by activating the Chk1-dependent pathway, while oxidative stress may be associated with eliciting toxicity in male reproductive cells.
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone and 0.5% 4-methyl-2-pentanone, S15, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required
Sigma-Aldrich
Ethyl alcohol, Pure, 160 proof, Excise Tax-free, Permit for use required
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 1% cyclohexane, A15 CYCLO1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Eosin Y disodium salt, certified by the Biological Stain Commission
Sigma-Aldrich
Eosin Y disodium salt, Dye content ≥85 %
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Eosin Y
Sigma-Aldrich
Eosin Y solution, 5 wt. % in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O